Skip to Content
Merck
Phthalic acid mono-2-ethylhexyl ester 97%

Phthalic acid mono-2-ethylhexyl ester

Phthalic acid mono-2-ethylhexyl ester 97%
Synonym(s):
mono-2-Ethylhexyl phthalate
Empirical Formula (Hill Notation):
C16H22O4
CAS Number:
Molecular Weight:
278.34

Products

Brand
Formula Weight
Physical Form
Purity
Functional Group
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing