
Phosphoric acid solution

Synonym(s):
Phosphoric acid, Orthophosphoric acid
Linear Formula:
H3PO4
CAS Number:
Molecular Weight:
98.00
Products
Product Category
Brand
Biological Source
Boiling Point (°C)
Formula Weight
Gene Alias
Melting Point (°C)
Physical Form
Agency
Quality Segment
Application
Markush Class
Markush Group
Organoleptic Class
Special Grade
Technique
Showing 1-5 of 5
Compare
Product Number
Description
Pricing