
Phosphate Standard for IC

Synonym(s):
Sodium phosphate dibasic solution, Disodium hydrogen phosphate solution, di-Sodium hydrogen phosphate solution
Linear Formula:
Na2HPO4
CAS Number:
Molecular Weight:
141.96
Products
Brand
Formula Weight
Product Line
Format
Quality Segment
Application
Reference Material Type
Special Grade
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing