Skip to Content
Merck
Phenyl chloroformate 99%

Phenyl chloroformate

Phenyl chloroformate 99%
Synonym(s):
Chloroformic acid phenyl ester
Linear Formula:
ClCOOC6H5
CAS Number:
Molecular Weight:
156.57
EC Number:

Products

Product Category
Brand
Boiling Point (°C)
Formula Weight
Physical Form
Purity
Quality Segment
Functional Group
Markush Class
Markush Group
Showing 1-2 of 2
Compare
Product Number
Description
Pricing