Skip to Content
AO
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Phenoxymethylpenicillin for system suitability
Synonym(s):
Phenoxymethylpenicillinic acid, Penicillin V
Empirical Formula (Hill Notation):
C
16
H
18
N
2
O
5
S
CAS Number:
87-08-1
Molecular Weight:
350.39
EC Number:
201-722-0
Products
Formula Weight
API Family
Reference Material Type
Filter Products
Showing 1-1 of 1
Compare
Product Number
Description
Pricing
Y0001868
Phenoxymethylpenicillin for system suitability,
European Pharmacopoeia (EP) Reference Standard
Expand
View Pricing