
Phenazine

Synonym(s):
Dibenzo[b,e]pyrazine, Phenazine
Empirical Formula (Hill Notation):
C12H8N2
CAS Number:
Molecular Weight:
180.21
Products
Brand
Formula Weight
Gene Alias
Melting Point (°C)
Physical Form
Purity
Quality Segment
Markush Class
Markush Group
Showing 1-2 of 2
Compare
Product Number
Description
Pricing