Skip to Content
AO
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Petroleum ether
Synonym(s):
Petroleum benzin
CAS Number:
101316-46-5
EC Number:
309-852-0
Products
Brand
Sigma-Aldrich (3)
Boiling Point (°C)
Physical Form
Agency
Quality Segment
Special Grade
Puriss (2)
Puriss P.A. (2)
Filter Products
Showing 1-3 of 3
Compare
Product Number
Description
Pricing
24587
Petroleum ether,
puriss., low boiling point hydrogen treated naphtha, meets analytical specification of DAB, bpmin. 75% 40-60 °C (min. 75%)
Expand
View Pricing
32247
Petroleum ether,
puriss. p.a., reag. Ph. Eur., high boiling, bp 50-70 °C
Expand
View Pricing
32248
Petroleum ether,
puriss. p.a., high boiling, bp 60-80 °C
Expand
View Pricing