Skip to Content
AO
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Permendur 49 alloy, Co49Fe49V2
Products
Manufacturer
Physical Form
Length (mm)
Filter Products
Showing 1-20 of 21
Page 1 of 2
Page 1 of 2
Compare
Product Number
Description
Pricing
GF14020131
Permendur 49 alloy, Co49Fe49V2,
sheet, 195 x 200mm, 0.35mm thickness, as rolled
Expand
View Pricing
GF03883945
Permendur 49 alloy, Co49Fe49V2,
sheet, 50 x 50mm, 0.25mm thickness, as rolled
Expand
View Pricing
GF25467903
Permendur 49 alloy, Co49Fe49V2,
foil, 100 x 100mm, 0.1mm thickness, as rolled
Expand
View Pricing
GF31309826
Permendur 49 alloy, Co49Fe49V2,
100mm rod, 25.4mm diameter, hot rolled (black)
Expand
View Pricing
GF53734631
Permendur 49 alloy, Co49Fe49V2,
50mm rod, 25.4mm diameter, hot rolled (black)
Expand
View Pricing
GF64047832
Permendur 49 alloy, Co49Fe49V2,
sheet, 300 x 300mm, 0.50mm thickness, as rolled
Expand
View Pricing
GF43852079
Permendur 49 alloy, Co49Fe49V2,
sheet, 50 x 50mm, 0.50mm thickness, as rolled
Expand
View Pricing
GF85493188
Permendur 49 alloy, Co49Fe49V2,
sheet, 100 x 100mm, 0.25mm thickness, as rolled
Expand
View Pricing
GF65381160
Permendur 49 alloy, Co49Fe49V2,
sheet, 150 x 150mm, 0.50mm thickness, as rolled
Expand
View Pricing
GF86648103
Permendur 49 alloy, Co49Fe49V2,
sheet, 150 x 150mm, 0.25mm thickness, as rolled
Expand
View Pricing
GF46199627
Permendur 49 alloy, Co49Fe49V2,
sheet, 60 x 100mm, 0.35mm thickness, as rolled
Expand
View Pricing
GF31013525
Permendur 49 alloy, Co49Fe49V2,
sheet, 180 x 500mm, 0.25mm thickness, as rolled
Expand
View Pricing
GF57695035
Permendur 49 alloy, Co49Fe49V2,
foil, 150 x 150mm, 0.1mm thickness, as rolled
Expand
View Pricing
GF66018157
Permendur 49 alloy, Co49Fe49V2,
foil, 25 x 25mm, 0.1mm thickness, as rolled
Expand
View Pricing
GF72140969
Permendur 49 alloy, Co49Fe49V2,
sheet, 100 x 100mm, 0.50mm thickness, as rolled
Expand
View Pricing
GF32567340
Permendur 49 alloy, Co49Fe49V2,
sheet, 100 x 195mm, 0.35mm thickness, as rolled
Expand
View Pricing
GF36940428
Permendur 49 alloy, Co49Fe49V2,
foil, 50 x 50mm, 0.1mm thickness, as rolled
Expand
View Pricing
GF91261829
Permendur 49 alloy, Co49Fe49V2,
foil, 300 x 300mm, 0.1mm thickness, as rolled
Expand
View Pricing
GF53199175
Permendur 49 alloy, Co49Fe49V2,
sheet, 95 x 100mm, 0.35mm thickness, as rolled
Expand
View Pricing
GF64205502
Permendur 49 alloy, Co49Fe49V2,
foil, 2m coil, 0.1mm thickness, as rolled
Expand
View Pricing
Page 1 of 2
Page 1 of 2