Skip to Content
AO
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Pentamidine isethionate salt
Synonym(s):
1,5-Bis(
p
-amidinophenoxy)pentane bis(2-hydroxyethanesulfonate salt), 4,4′-(Pentamethylenedioxy)dibenzamidine bis(2-hydroxyethanesulfonate)
Linear Formula:
CH
2
[CH
2
CH
2
OC
6
H
4
C(=NH)NH
2
]
2
·2HOCH
2
CH
2
SO
3
H
CAS Number:
140-64-7
Molecular Weight:
592.68
EC Number:
205-424-1
Products
Product Category
bioactive small molecules (1)
Brand
Sigma-Aldrich (1)
Formula Weight
Gene Alias
Melting Point (°C)
Physical Form
Activity Spectrum
Research Area
Quality Segment
Mode of Action
Filter Products
Showing 1-1 of 1
Compare
Product Number
Description
Pricing
P0547
Pentamidine isethionate salt,
powder
Expand
View Pricing