Skip to Content
AO
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
PCL-PEG-PCL diacrylate
Linear Formula:
C
2
H
3
O
2
(C
6
H
10
O
2
)
x
(C
2
H
4
O)
y
(OC
6
H
10
O)
z
OC
2
H
3
Products
Brand
Sigma-Aldrich (9)
Color
Feature
Physical Form
Quality Segment
Functional Group
Filter Products
Showing 1-9 of 9
Compare
Product Number
Description
Pricing
929611
PCL-PEG-PCL diacrylate,
PCL average M
n
800, PEG average M
n
4000
Expand
View Pricing
929786
PCL-PEG-PCL diacrylate,
PCL average M
n
200, PEG average M
n
1000
Expand
View Pricing
929573
PCL-PEG-PCL diacrylate,
average M
n
1000-10000-1000, Poly(caprolactone)-
b
-poly(ethylene glycol)-
b
-poly(caprolactone) diacrylate
Expand
View Pricing
929581
PCL-PEG-PCL diacrylate,
average M
n
2000-20000-2000, poly(caprolactone)-
b
-poly(ethylene glycol)-
b
-poly(caprolactone) diacrylate
Expand
View Pricing
929794
PCL-PEG-PCL diacrylate,
average M
n
(200-2000-200), poly(caprolactone)-
b
-poly(ethylene glycol)-
b
-poly(caprolactone) diacrylate
Expand
View Pricing
929603
PCL-PEG-PCL diacrylate,
average M
n
1000-6000-1000, Poly(caprolactone)-
b
-poly(ethylene glycol)-
b
-poly(caprolactone) diacrylate
Expand
View Pricing
929824
PCL-PEG-PCL diacrylate,
average M
n
(600-3400-600), poly(caprolactone)-
b
-poly(ethylene glycol)-
b
-poly(caprolactone) diacrylate
Expand
View Pricing
929832
PCL-PEG-PCL diacrylate,
average M
n
(600-6000-600), poly(caprolactone)-
b
-poly(ethylene glycol)-
b
-poly(caprolactone) diacrylate
Expand
View Pricing
929816
PCL-PEG-PCL diacrylate,
average M
n
(300-3400-300), poly(caprolactone)-
b
-poly(ethylene glycol)-
b
-poly(caprolactone) diacrylate
Expand
View Pricing