Skip to Content
AO
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Paracetamol
Synonym(s):
Acetaminophen, 4′-Hydroxyacetanilide, 4-Acetamidophenol,
N
-(4-Hydroxyphenyl)acetamide,
N
-Acetyl-4-aminophenol, APAP, Paracetamol
Linear Formula:
CH
3
CONHC
6
H
4
OH
CAS Number:
103-90-2
Molecular Weight:
151.16
EC Number:
203-157-5
Products
Formula Weight
Gene Alias
Manufacturer
Melting Point (°C)
Physical Form
Format
Application
Markush Class
Markush Group
API Family
Reference Material Type
Special Grade
pharmaceutical primary standard (2)
Technique
Filter Products
Showing 1-2 of 2
Compare
Product Number
Description
Pricing
BP371
Paracetamol,
British Pharmacopoeia (BP) Reference Standard
Expand
View Pricing
P0300000
Paracetamol,
European Pharmacopoeia (EP) Reference Standard
Expand
View Pricing