
Paclitaxel semi-synthetic for system suitability

Synonym(s):
Paclitaxel
Empirical Formula (Hill Notation):
C47H51NO14
CAS Number:
Molecular Weight:
853.91
Products
Formula Weight
Gene Alias
Manufacturer
Melting Point (°C)
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing