
Orphenadrine Related Compound F

Synonym(s):
4-Methyldiphenhydramine citrate salt, N,N-Dimethyl-2-[phenyl(p-tolyl)methoxy]ethanamine citrate, p-Methyldiphenhydramine citrate salt
Empirical Formula (Hill Notation):
C18H23NO · C6H8O7
CAS Number:
Molecular Weight:
461.50
Products
Formula Weight
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing