Skip to Content
AO
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Orcinol monohydrate
Synonym(s):
3,5-Dihydroxytoluene, 5-Methylresorcinol
Linear Formula:
CH
3
C
6
H
3
-1,3-(OH)
2
· H
2
O
CAS Number:
6153-39-5
Molecular Weight:
142.15
EC Number:
207-984-2
Products
Product Category
stains (1)
Brand
Sigma-Aldrich (1)
Biological Source
Boiling Point (°C)
Color
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Application
Technique
Filter Products
Showing 1-1 of 1
Compare
Product Number
Description
Pricing
O1875
Orcinol monohydrate,
≥99% purity (HPLC), powder
Expand
View Pricing