Skip to Content
Merck
o-Phenylenediamine dihydrochloride tablet, 1 mg substrate per tablet

o-Phenylenediamine dihydrochloride

o-Phenylenediamine dihydrochloride tablet, 1 mg substrate per tablet
Linear Formula:
C6H4(NH2)2·2HCl
CAS Number:
Molecular Weight:
181.06

Products

Product Category
Brand
Color
Formula Weight
Melting Point (°C)
Physical Form
Quality Segment
Markush Class
Markush Group