
O,O′-Bis(2-aminopropyl) polypropylene glycol-block-polyethylene glycol-block-polypropylene glycol

Linear Formula:
CH3CH(NH2)CH2[OCH(CH3)CH2]l(OCH2CH2)m[OCH2CH(CH3)]nNH2
CAS Number:
Products
Melting Point (°C)
pH Value
Physical Form
Polymer Composition
Polymer Type