Skip to Content
Merck
O-Methylisourea hemisulfate salt 99%

O-Methylisourea hemisulfate salt

O-Methylisourea hemisulfate salt 99%
Synonym(s):
OMI®
Linear Formula:
H2NC(OCH3)=NH · 1/2H2SO4
CAS Number:
Molecular Weight:
123.12
EC Number:

Products

Product Category
Brand
Formula Weight
Gene Alias
Melting Point (°C)
Physical Form
Purity
Functional Group
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing