
Olmesartan medoxomil for system suitability

Synonym(s):
Olmesartan medoxomil
Empirical Formula (Hill Notation):
C29H30N6O6
CAS Number:
Molecular Weight:
558.59
Products
Formula Weight
Gene Alias
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing