Skip to Content
AO
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Octyl acetate
Synonym(s):
Acetic acid octyl ester
Linear Formula:
CH
3
CO
2
(CH
2
)
7
CH
3
CAS Number:
112-14-1
Molecular Weight:
172.26
EC Number:
203-939-6
Products
Product Category
aroma chemicals (2)
building blocks (1)
Brand
Sigma-Aldrich (3)
Biological Source
Boiling Point (°C)
Formula Weight
Purity
Agency
Quality Segment
Application
Markush Class
Markush Group
Organoleptic Class
Special Grade
Food Grade (2)
Fragrance grade (1)
Halal (1)
Kosher (1)
Natural (1)
Filter Products
Showing 1-3 of 3
Compare
Product Number
Description
Pricing
O5500
Octyl acetate,
≥99%
Expand
View Pricing
W280607
Octyl acetate,
≥98%, FCC, FG
Expand
View Pricing
W280615
Octyl acetate,
natural, ≥98%, FCC, FG
Expand
View Pricing