Skip to Content
Merck
N-tert-Butylmaleimide 97%

N-tert-Butylmaleimide

N-tert-Butylmaleimide 97%
Empirical Formula (Hill Notation):
C8H11NO2
CAS Number:
Molecular Weight:
153.18

Products

Product Category
Brand
Boiling Point (°C)
Formula Weight
Physical Form
Purity
Functional Group
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing