Skip to Content
Merck
Novaluron PESTANAL®, analytical standard

Novaluron

Novaluron PESTANAL®, analytical standard
Synonym(s):
1-[3-Chloro-4-(1,1,2-trifluoro-2-trifluoromethoxyethoxy)phenyl]-3-(2,6-difluorobenzoyl)urea
Empirical Formula (Hill Notation):
C17H9ClF8N2O4
CAS Number:
Molecular Weight:
492.70

Products

Product Category
Brand
Formula Weight
Manufacturer
Product Line
Format
Quality Segment
Application
Reference Material Type
Special Grade
Technique