
Norgestimate for system suitability

Synonym(s):
Norgestimate, (17α)-17-(Acetyloxy)-13-ethyl-18,19-dinorpregn-4-en-20-yn-3-one 3-oxime, Dexnorgestrel acetime
Empirical Formula (Hill Notation):
C23H31NO3
CAS Number:
Molecular Weight:
369.50
Products
Formula Weight
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing