
Nonylphenol

Empirical Formula (Hill Notation):
C15H24O
Products
Product Category
Brand
Formula Weight
Gene Alias
Product Line
Format
Quality Segment
Application
Reference Material Type
Special Grade
Technique
Showing 1-2 of 2
Compare
Product Number
Description
Pricing