Skip to Content
Merck
Nonyl alcohol ≥98%, FCC

Nonyl alcohol

Nonyl alcohol ≥98%, FCC
Synonym(s):
1-Nonanol, Alcohol C9, Nonyl alcohol
Linear Formula:
CH3(CH2)8OH
CAS Number:
Molecular Weight:
144.25
EC Number:

Products

Product Category
Biological Source
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Purity
Agency
Quality Segment
Application
Markush Class
Markush Group
Organoleptic Class
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing