N,N-Dimethyl-L-phenylalanine
Linear Formula:
C6H5CH2CH[N(CH3)2]CO2H
CAS Number:
Molecular Weight:
193.24
Products
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Application
Markush Class
Markush Group
Optical Isomer
Amino Acid
Amino Acid Type
Reaction Type
Showing 1-1 of 1
Compare
Product Number
Description
Pricing