Skip to Content
AO
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
N
,
N
-Dimethylformamide dipropyl acetal
Synonym(s):
1,1-Dipropoxy-
N
,
N
-dimethylmethylamine, 1,1-Dipropoxytrimethylamine
Linear Formula:
(CH
3
)
2
NCH(OCH
2
CH
2
CH
3
)
2
CAS Number:
6006-65-1
Molecular Weight:
175.27
EC Number:
227-855-4
Products
Product Category
building blocks (1)
Brand
Supelco (1)
Boiling Point (°C)
Formula Weight
Physical Form
Quality Segment
Markush Class
Markush Group
Reaction Type
Reagent Type
Technique
Filter Products
Showing 1-1 of 1
Compare
Product Number
Description
Pricing
394998
N
,
N
-Dimethylformamide dipropyl acetal,
for GC derivatization, LiChropur
™
Expand
View Pricing