Skip to Content
Merck
N,N-Dimethylbenzylamine ≥99%

N,N-Dimethylbenzylamine

N,N-Dimethylbenzylamine ≥99%
Synonym(s):
N-Benzyldimethylamine, BDMA, DMBA, N,N-Dimethylbenzylamine
Linear Formula:
C6H5CH2N(CH3)2
CAS Number:
Molecular Weight:
135.21

Products

Product Category
Brand
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Functional Group
Markush Class
Markush Group
Showing 1-2 of 2
Compare
Product Number
Description
Pricing