Skip to Content
Merck
N,N-Dimethylacetoacetamide solution 80% in H2O

N,N-Dimethylacetoacetamide solution

N,N-Dimethylacetoacetamide solution 80% in H2O
Synonym(s):
N,N-Dimethyl-3-oxobutyramide
Linear Formula:
CH3COCH2CON(CH3)2
CAS Number:
Molecular Weight:
129.16

Products

Product Category
Boiling Point (°C)
Formula Weight
Functional Group
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing