
Nitrilotriacetic acid disodium salt

Synonym(s):
N,N-Bis(carboxymethyl)glycine, NTA, Triglycollamic acid
Linear Formula:
C6H7NO6Na2
Products
Product Category
Brand
Formula Weight
Physical Form
Purity
Quality Segment
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing