Skip to Content
Merck
Niobium rod, 200mm, diameter 20.0mm, annealed, 99.9%

Niobium

Niobium rod, 200mm, diameter 20.0mm, annealed, 99.9%
Synonym(s):
Columbium, Niobium
Empirical Formula (Hill Notation):
Nb
CAS Number:
Molecular Weight:
92.91

Products

Brand
Boiling Point (°C)
Formula Weight
Manufacturer
Melting Point (°C)
Physical Form
Diameter (mm)
Length (m)
Length (mm)
Element
Quality Segment
Showing 1-20 of 201
Page 1 of 11
Compare
Product Number
Description
Pricing
Page 1 of 11