Skip to Content
AO
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Nickel silver alloy, Cu45Zn43Ni10Pb2
Products
Manufacturer
Physical Form
Diameter (mm)
Length (mm)
Filter Products
Showing 1-4 of 4
Compare
Product Number
Description
Pricing
GF65974203
Nickel silver alloy, Cu45Zn43Ni10Pb2,
500mm rod, 6.35mm diameter
Expand
View Pricing
GF74609185
Nickel silver alloy, Cu45Zn43Ni10Pb2,
100mm rod, 6.35mm diameter
Expand
View Pricing
GF06834517
Nickel silver alloy, Cu45Zn43Ni10Pb2,
1000mm rod, 6.35mm diameter
Expand
View Pricing
GF73182450
Nickel silver alloy, Cu45Zn43Ni10Pb2,
200mm rod, 6.35mm diameter
Expand
View Pricing