Skip to Content
Merck
Neutral red (C.I. 50040)

Neutral red (C.I. 50040)

Neutral red (C.I. 50040)
Synonym(s):
Neutral red (C.I. 50040)
Empirical Formula (Hill Notation):
C15H17ClN4
CAS Number:
Molecular Weight:
288.78

Products

Product Category
Brand
Formula Weight
Physical Form
Chemical Composition
Quality Segment
Application
Showing 1-2 of 2
Compare
Product Number
Description
Pricing