
Nepafenac

Synonym(s):
2-Amino-3-benzoyl-benzeneacetamide, AHR 9434, AL 6515
Empirical Formula (Hill Notation):
C15H14N2O2
CAS Number:
Molecular Weight:
254.28
Products
Product Category
Brand
Color
Formula Weight
Gene Alias
Manufacturer
Physical Form
Purity
Format
Research Area
Quality Segment
Application
API Family
Reference Material Type
Showing 1-2 of 2
Compare
Product Number
Description
Pricing