Skip to Content
Merck
N-(Benzyloxycarbonyloxy)succinimide 98%

N-(Benzyloxycarbonyloxy)succinimide

N-(Benzyloxycarbonyloxy)succinimide 98%
Synonym(s):
Benzyl N-succinimidyl carbonate, Z-OSu
Empirical Formula (Hill Notation):
C12H11NO5
CAS Number:
Molecular Weight:
249.22
EC Number:

Products

Brand
Formula Weight
Melting Point (°C)
Purity
Functional Group
Application
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing