
Naproxen (racemic)

Synonym(s):
(±)-Naproxen, 2-(6-Methoxy-2-naphthyl)propionic acid
Linear Formula:
CH3OC10H6CH(CH3)CO2H
CAS Number:
Molecular Weight:
230.26
Products
Formula Weight
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing