
Naftifine hydrochloride

Empirical Formula (Hill Notation):
C21H21N · HCl
CAS Number:
Molecular Weight:
323.86
Products
Product Category
Brand
Formula Weight
Manufacturer
Physical Form
Purity
Activity Spectrum
Format
Quality Segment
Application
Mode of Action
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing