Skip to Content
AO
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
MTC
™
Bio SureSeal S
™
Microcentrifuge Tube
Products
Product Category
tubes (14)
Manufacturer
Capacity (mL)
Color
Package Size
Sterility
Filter Products
Showing 1-14 of 14
Compare
Product Number
Description
Pricing
MTCC200050
MTC
™
Bio SureSeal S
™
Microcentrifuge Tube,
capacity 1.5 mL, sterile, clear, pack of 500 ea (10 x packs 50ea)
Expand
View Pricing
MTCC2002
MTC
™
Bio SureSeal S
™
Microcentrifuge Tube,
capacity 2.0 mL, sterile, clear, pack of 500 ea
Expand
View Pricing
MTCC2007
MTC
™
Bio SureSeal S
™
Microcentrifuge Tube,
capacity 0.5 mL, sterile, clear, pack of 500 ea
Expand
View Pricing
MTCC2000
MTC
™
Bio SureSeal S
™
Microcentrifuge Tube,
capacity 1.5 mL, sterile, clear, pack of 500 ea
Expand
View Pricing
MTCC2007AST
MTC
™
Bio SureSeal S
™
Microcentrifuge Tube,
capacity 0.5 mL, sterile, assorted colors, pack of 500 ea
Expand
View Pricing
MTCC2000OB
MTC
™
Bio SureSeal S
™
Microcentrifuge Tube,
capacity 1.5 mL, sterile, black, pack of 500 ea
Expand
View Pricing
MTCC200050AST
MTC
™
Bio SureSeal S
™
Microcentrifuge Tube,
capacity 1.5 mL, sterile, assorted colors, pack of 500 ea (10 x bags 50 ea, single colour per pack)
Expand
View Pricing
MTCC2000AST
MTC
™
Bio SureSeal S
™
Microcentrifuge Tube,
capacity 1.5 mL, sterile, assorted colors, pack of 500 ea
Expand
View Pricing
MTCC2000Y
MTC
™
Bio SureSeal S
™
Microcentrifuge Tube,
capacity 1.5 mL, sterile, yellow, pack of 500 ea
Expand
View Pricing
MTCC2000O
MTC
™
Bio SureSeal S
™
Microcentrifuge Tube,
capacity 1.5 mL, sterile, orange, pack of 500 ea
Expand
View Pricing
MTCC2000P
MTC
™
Bio SureSeal S
™
Microcentrifuge Tube,
capacity 1.5 mL, sterile, purple, pack of 500 ea
Expand
View Pricing
MTCC2000R
MTC
™
Bio SureSeal S
™
Microcentrifuge Tube,
capacity 1.5 mL, sterile, red, pack of 500 ea
Expand
View Pricing
MTCC2000B
MTC
™
Bio SureSeal S
™
Microcentrifuge Tube,
capacity 1.5 mL, sterile, blue, pack of 500 ea
Expand
View Pricing
MTCC2000G
MTC
™
Bio SureSeal S
™
Microcentrifuge Tube,
capacity 1.5 mL, sterile, green, pack of 500 ea
Expand
View Pricing