Skip to Content
AO
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
MOPS
Synonym(s):
3-(N-Morpholino)propanesulfonic acid, 4-Morpholinepropanesulfonic acid, MOPS, Free Acid, ULTROL
®
Grade, 3-Morpholinopropane sulfonic acid
Empirical Formula (Hill Notation):
C
7
H
15
NO
4
S
CAS Number:
1132-61-2
Molecular Weight:
209.26
EC Number:
214-478-5
Products
Product Category
buffers (6)
Brand
Millipore (2)
Sigma-Aldrich (1)
Color
Feature
Formula Weight
pH Value
Physical Form
pK Value
Product Line
Purity
Research Area
Quality Segment
Application
Special Grade
Anhydrous (1)
BioPerformance Certified (1)
Technique
Filter Products
Showing 1-7 of 7
Compare
Product Number
Description
Pricing
M1254
MOPS,
≥99.5% (titration), crystalline powder
Expand
View Pricing
M3183
MOPS,
≥99.5% (titration), crystalline powder, BioPerformance Certified, suitable for cell culture
Expand
View Pricing
M5162
MOPS,
BioXtra, ≥99.5% (titration)
Expand
View Pricing
475898
MOPS,
≥99% (titration), solid, Ultrol
®
Grade
Expand
View Pricing
1.06129
MOPS,
solid, buffer substance MPS
Expand
View Pricing
RDD003
MOPS,
≥99.5%, crystalline powder, anhydrous, Redi-Dri
™
Expand
View Pricing
69947
MOPS,
≥99.5% (titration), powder or crystals, for molecular biology, BioUltra
Expand
View Pricing