
Monoclonal Anti-β-Actin antibody produced in mouse

Synonym(s):
Monoclonal Anti-β-Actin
Products
Product Category
Brand
Biological Source
Clonality
Gene Alias
Isotype
Physical Form
Antibody Form
Research Area
Conjugate
Quality Segment
Application
Post Translational Modification
Non-reactive Species
Species Reactivity
Technique
Showing 1-3 of 3
Compare
Product Number
Description
Pricing