Skip to Content
AO
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Molymod
®
Atoms and Bonds
Products
Color
Manufacturer
Diameter (mm)
Filter Products
Showing 1-7 of 7
Compare
Product Number
Description
Pricing
Z543950
Molymod
®
Atoms and Bonds,
Fluorine, no. of holes, 1, diam. 17 mm
Expand
View Pricing
Z544167
Molymod
®
Atoms and Bonds,
Carbon, no. of holes, 4, diam. 23 mm, angle, 109.5
Expand
View Pricing
Z543896
Molymod
®
Atoms and Bonds,
Hydrogen, no. of holes, 1
Expand
View Pricing
Z544094
Molymod
®
Atoms and Bonds,
Metal, no. of holes, 2, diam. 23 mm, angle, 105.00
Expand
View Pricing
Z544108
Molymod
®
Atoms and Bonds,
Nitrogen, no. of holes, 2, diam. 23 mm, angle, 105.00
Expand
View Pricing
Z544221
Molymod
®
Atoms and Bonds,
Halogen, no. of holes, 4, diam. 23 mm, angle, 109.50
Expand
View Pricing
Z544191
Molymod
®
Atoms and Bonds,
Sulfur, no. of holes, 4, diam. 23 mm, angle, 109.50
Expand
View Pricing