Skip to Content
Merck
Mitoxantrone dihydrochloride ≥97% (HPLC)

Mitoxantrone dihydrochloride

Mitoxantrone dihydrochloride ≥97% (HPLC)
Synonym(s):
1,4-Dihydroxy-5,8-bis[[2-[(2-hydroxyethyl)amino]ethyl]amino]-9,10-anthracenedione dihydrochloride
Empirical Formula (Hill Notation):
C22H28N4O6 · 2HCl
CAS Number:
Molecular Weight:
517.40
EC Number:

Products

Product Category
Brand
Formula Weight
Gene Alias
Purity
Showing 1-1 of 1
Compare
Product Number
Description
Pricing