
Micro particles, magnetic, streptavidin coated

Synonym(s):
Polystyrene beads, magnetic, streptavidin coated
Products
Product Category
Brand
Physical Form
Format
Chemical Composition
Quality Segment
Application
Particle Size (µm)
Reference Material Type
Special Grade
Showing 1-2 of 2