Skip to Content
AO
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Micro particles based on melamine resin
Synonym(s):
Melamine-formaldehyde resin, Size standards
Products
Product Category
resins (1)
Brand
Sigma-Aldrich (1)
Supelco (2)
Physical Form
Format
Quality Segment
Particle Size (µm)
Reference Material Type
Special Grade
analytical standard (2)
Filter Products
Showing 1-3 of 3
Compare
Product Number
Description
Pricing
96062
Micro particles based on melamine resin,
analytical standard, size: 10 μm
Expand
View Pricing
90637
Micro particles based on melamine resin,
size: 2 μm
Expand
View Pricing
95523
Micro particles based on melamine resin,
analytical standard, size: 8 μm
Expand
View Pricing