Skip to Content
AO
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Methyltrichlorosilane
Synonym(s):
Trichloro(methyl)silane
Linear Formula:
CH
3
SiCl
3
CAS Number:
75-79-6
Molecular Weight:
149.48
EC Number:
200-902-6
Products
Brand
Sigma-Aldrich (3)
Boiling Point (°C)
Formula Weight
Physical Form
Purity
Quality Segment
Filter Products
Showing 1-3 of 3
Compare
Product Number
Description
Pricing
M85301
Methyltrichlorosilane,
99%
Expand
View Pricing
440299
Methyltrichlorosilane,
≥96%
Expand
View Pricing
679208
Methyltrichlorosilane,
deposition grade, ≥98% (GC), ≥99.99% (as metals)
Expand
View Pricing