Skip to Content
Merck
Methyltestosterone certified reference material, pharmaceutical secondary standard

Methyltestosterone

Methyltestosterone certified reference material, pharmaceutical secondary standard
Synonym(s):
17α-Methyltestosterone, 17α-Methyl-4-androsten-17β-ol-3-one, 17β-Hydroxy-17α-methyl-4-androsten-3-one, Mesterone, Methyltestosterone
Empirical Formula (Hill Notation):
C20H30O2
CAS Number:
Molecular Weight:
302.45

Products

Brand
Formula Weight
Melting Point (°C)
Agency
Quality Segment
Application
Markush Class
Markush Group
Package Size
Special Grade
Showing 1-1 of 1