Methyl hexanoate
Synonym(s):
Caproic acid methyl ester, Methyl caproate, Hexanoic acid methyl ester, Methyl hexanoate
Linear Formula:
CH3(CH2)4COOCH3
CAS Number:
Molecular Weight:
130.18
Products
Product Category
Brand
Sustainability
Biological Source
Boiling Point (°C)
Feature
Formula Weight
Melting Point (°C)
Physical Form
Purity
Agency
Format
Quality Segment
Functional Group
Application
Markush Class
Markush Group
Organoleptic Class
Reference Material Type
Special Grade
Technique
Showing 1-5 of 5