Skip to Content
Merck
Methyl Blue for microscopy (Bot., Hist.), indicator (pH 9.4-14.0)

Methyl Blue

Methyl Blue for microscopy (Bot., Hist.), indicator (pH 9.4-14.0)
Synonym(s):
Acid blue 93, Aniline blue water soluble, Cotton blue, Poirriers blue, Water blue
Empirical Formula (Hill Notation):
C37H27N3Na2O9S3
CAS Number:
Molecular Weight:
799.80
EC Number:

Products

Product Category
Brand
Formula Weight
Physical Form
Quality Segment
Application
Technique