
Methyl 2-oxocyclopentanecarboxylate

Synonym(s):
2-Methoxycarbonylcyclopentanone, Methyl cyclopentanone-2-carboxylate
Linear Formula:
(O=)C5H7CO2CH3
Products
Product Category
Brand
Boiling Point (°C)
Formula Weight
Purity
Functional Group
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing