Skip to Content
Merck
Methyl 2-methyl-3-nitrobenzoate 97%

Methyl 2-methyl-3-nitrobenzoate

Methyl 2-methyl-3-nitrobenzoate 97%
Synonym(s):
Methyl 3-nitro-o-toluate
Linear Formula:
CH3C6H3(NO2)CO2CH3
CAS Number:
Molecular Weight:
195.17

Products

Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Functional Group
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing