Methyl 1-(butylcarbamoyl)-2-benzimidazolecarbamate
Synonym(s):
Benomyl, NSC 263489
Empirical Formula (Hill Notation):
C14H18N4O3
Products
Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Functional Group
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing